Få mere at vide
STEMCELL Technologies EasySep™ Human Whole Blood and Bone Marrow CD138 Positive Selection Kit II
Beskrivelse
- The EasySep™ Human Whole Blood and Bone Marrow CD138 Positive Selection Kit is designed to isolate CD138+ (syndecan-1) cells from whole blood and bone marrow by positive selection
- Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD138 and dextran-coated magnetic particles
- The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Cells of interest remain in the tube while unwanted cells are poured off
- The CD138 antigen is expressed on normal and malignant plasma cells (but not mature B cells)
- This product replaces the EasySep™ Human Whole Blood and Bone Marrow CD138 Positive Selection Kit (Catalog #18387) for even faster cell isolations
Tekniske data
Tekniske data
| Celletype | Myeloma, Plasma |
| Til brug med (applikation) | ELISA |
| Mængde | 60 mL Whole Blood and Bone Marrow |