Learn More
STEMCELL Technologies EasySep™ Human Resting CD4+ T Cell Isolation Kit
Immunomagnetic isolation of untouched human resting CD4+ T cells
Brand: STEMCELL Technologies 17962
This item is not returnable.
View return policy
Description
- The EasySep™ Human Resting CD4+ T Cell Isolation Kit is designed to isolate resting CD4+ T cells from fresh or previously frozen human peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection
- The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles
- The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube
Specifications
T Cells, T Cells, CD4+ | |
Cell Isolation | |
1 x 109 Cells |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
STEMCELL Technologies EasySep™ Human Resting CD4+ T Cell Isolation Kit
Spot an opportunity for improvement?Share a Content Correction