Få mere at vide
STEMCELL Technologies EasySep™ Human PE Positive Selection Kit II
Artikelnummer. 17187741
Encompass
Mængde:
1 x 109 cells
5 x 109 cells
Pakningsstørrelse:
1 sæt
Artikelnummer. 17187741
Brand: STEMCELL Technologies 17694
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Immunomagnetic positive selection of human cells labeled with PE-conjugated antibodies
  • The EasySep™ Human PE Positive Selection Kit II is designed to isolate cells that are labeled with PE (phycoerythrin)-conjugated antibodies by positive selection
  • Desired cells are targeted with antibody complexes recognizing PE and dextran-coated magnetic particles
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Cells of interest remain in the tube while unwanted cells are poured off
  • This product replaces the EasySep™ Human PE Positive Selection Kit (Catalog #18551)
Tekniske data
Celletype B Cells, Dendritic Cells, Granulocytes and Subsets, Hematopoietic Stem and Progenitor Cells, Macrophages, Marrow Stromal Cells, Mesenchymal Stem and Progenitor Cells, Monocytes, Mononuclear Cells, Myeloid Cells, NK Cells, Other, Plasma, T Cells
Til brug med (applikation) Cell Isolation
Mængde 5 x 109 cells
Korrektion af produktindhold
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.