Learn More
STEMCELL Technologies EasySep™ Human Naïve CD8+ T Cell Isolation Kit II
Immunomagnetic isolation of untouched human naïve CD8+ T cells
Brand: STEMCELL Technologies 17968
Description
- The EasySep™ Human Naïve CD8+ T Cell Isolation Kit II is designed to isolate naïve CD8+ T cells from fresh peripheral blood mononuclear cells by negative selection
- The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles
- The magnetically-labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube
- This product replaces the EasySep™ Human Naïve CD8+ T Cell Enrichment Kit (Catalog #19158) for even faster cell isolations
Specifications
T Cells, T Cells, CD8+ | |
Cell Isolation | |
1 x 109 cells |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.