missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EAF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA564418
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: GSDDDSSSSG GEDNGPASPP QPSHQQPYNS RPAVANGTSR PQGSNQLMNT LRN Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 91%.
EAF1 Polyclonal antibody specifically detects EAF1 in Human samples. It is validated for Immunohistochemistry.Specifications
EAF1 | |
Polyclonal | |
Unconjugated | |
EAF1 | |
4933403C17Rik; AI666536; EAF1; ELL (eleven nineteen lysine-rich leukemia gene)-associated factor 1; ELL associated factor 1; ELL-associated factor 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
85403 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96JC9 | |
EAF1 | |
Recombinant protein corresponding to Human EAF1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only