Få mere at vide
(E/Z)-Geranylacetone, TRC
Artikelnummer. 30240854
Skift visning
Mængde:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240854 5 g
30240541 10 g
30240701 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30240854 Leverandør Toronto Research Chemicals Leverandørnr. TRCG3677785G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 689-67-8
Molekylær formel C13H22O
Molekylvægt (g/mol) 194.31
Synonym Dihydropseudoionone, 6,10-Dimethyl-5,9-undecadien-2-one, 2,6-Dimethylundeca-2,6-dien-10-one, 6,10-Dimethyl-5,9-undec-2-one, 6,10-Dimethyl-5,9-undecadiene-2-one;
SMIL CC(=CCC\C(=C\CCC(=O)C)\C)C
Tekniske data
Kemisk navn eller materiale (E/Z)-Geranylacetone
Mængde 5 g
InChI formel InChI=1S/C13H22O/c1-11(2)7-5-8-12(3)9-6-10-13(4)14/h7,9H,5-6,8,10H2,1-4H3/b12-9+