Få mere at vide
(E/Z)-Chlorprothixene-d6 Hydrochloride(Mixture), TRC
Artikelnummer. 30350565
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30350565 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30350565 Leverandør Toronto Research Chemicals Leverandørnr. TRCC42485350MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (E/Z)-Chlorprothixene-d6 Hydrochloride
CAS 1246832-91-6
Molekylær formel C18H13D6Cl2NS
Mængde 50 mg
Synonym (3E/Z)-3-(2-Chloro-9H-thioxanthen-9-ylidene)-N,N-(dimethyl-d6)-1-propanamine Hydrochloride, cis/trans-Chlorprothixene-d6 Hydrochloride
InChI formel InChI=1S/C18H18ClNS.ClH/c1-20(2)11-5-7-14-15-6-3-4-8-17(15)21-18-10-9-13(19)12-16(14)18;/h3-4,6-10,12H,5,11H2,1-2H3;1H/b14-7+;/i1D3,2D3;
SMIL ClC1=CC2=C(C=C1)SC3=CC=CC=C3/C2=C\CCN(C([2H])([2H])[2H])C([2H])([2H])[2H].Cl
IUPAC navn (3E)-3-(2-chlorothioxanthen-9-ylidene)-N,N-bis(trideuteriomethyl)propan-1-amine;hydrochloride
Molekylvægt (g/mol) 358.36
Formel vægt 357.0992