Få mere at vide
(E/Z)-4-Carboxymethyl-5-(4-fluorophenyl)-2-methyl-pent-4-en-3-one, TRC
Artikelnummer. 30361217
Skift visning
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30361217 50 mg
30349011 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30361217 Leverandør Toronto Research Chemicals Leverandørnr. TRCC17875050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 122549-26-2
Molekylær formel C14H15FO3
Molekylvægt (g/mol) 250.27
Synonym 2-[(4-Fluorophenyl)methylene]-4-methyl-3-oxo-pentanoic Acid Methyl Ester
IUPAC navn methyl (2E)-2-[(4-fluorophenyl)methylidene]-4-methyl-3-oxopentanoate
SMIL CC(C)C(=O)\C(=C\c1ccc(F)cc1)\CC(=O)O
Tekniske data
Kemisk navn eller materiale (E/Z)-4-Carboxymethyl-5-(4-fluorophenyl)-2-methyl-pent-4-en-3-one
Mængde 50 mg
InChI formel InChI=1S/C14H15FO3/c1-9(2)13(16)12(14(17)18-3)8-10-4-6-11(15)7-5-10/h4-9H,1-3H3/b12-8+