Få mere at vide
(E,Z)-3-(Dibenzo[b,e]oxepin-11(6H)-ylidene)-N-methylpropan-1-amine ((E,Z)-Desmethyldoxepin), Mikromol™
Beskrivelse
(E,Z)-3-(Dibenzo[b,e]oxepin-11(6H)-ylidene)-N-methylpropan-1-amine ((E,Z)-Desmethyldoxepin)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (E,Z)-Desmethyldoxepin |
| CAS | 1225-56-5 |
| Analyt- eller komponentnavne | (E,Z)-Desmethyldoxepin |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C18 H19 N O |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 1-Propanamine, 3-dibenz[b,e]oxepin-11(6H)-ylidene-N-methyl-, Dibenz[b,e]oxepin-Δ11(6H),γ-propylamine, N-methyl- (7CI,8CI), 3-Dibenz[b,e]oxepin-11(6H)-ylidene-N-methyl-1-propanamine, Dibenz[b,e]oxepin, 1-propanamine deriv., Demethyldoxepin, Desdoxepin, Desmethyl doxepin, Desmethyldoxepine, Monodesmethyldoxepin, N-Desmethyldoxepin, Nordoxepin, (E,Z)-3-(Dibenzo[b,e]oxepin-11(6H)-ylidene)-N-methylpropan-1-amine, Doxepin Hydrochloride Imp. C (EP) as E/Z-Mixture, (E,Z)-Desmethyldoxepin, Desmethyldoxepin (cis/trans) |
| InChI formel | InChI=1S/C18H19NO/c1-19-12-6-10-16-15-8-3-2-7-14(15)13-20-18-11-5-4-9-17(16)18/h2-5,7-11,19H,6,12-13H2,1H3 |
| SMIL | CNCCC=C1c2ccccc2COc3ccccc13 |
| IUPAC navn | 3-(6H-benzo[c][1]benzoxepin-11-ylidene)-N-methylpropan-1-amine |
| Vis mere |