Få mere at vide
(E,Z)-2-Propyl-2-pentenoic Acid, TRC
Artikelnummer. 30420838
Skift visning
Mængde:
1 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30420838 100 mg
30412756 1 mg
30415133 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30420838 Leverandør Toronto Research Chemicals Leverandørnr. TRCP838300100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 60218-41-9
Molekylær formel C8 H14 O2
Molekylvægt (g/mol) 142.2
Synonym 2-Pentenoic acid, 2-propyl-, 2-Propyl-2-pentenoic acid, 2-Propyl-2-pentenoate, 2-Propylpenten-2-oic acid, (E,Z)-2-Propyl-2-pentenoic acid
IUPAC navn (E)-2-propylpent-2-enoic acid
SMIL CCC\C(=C/CC)\C(=O)O
Tekniske data
Kemisk navn eller materiale (E,Z) 2-Propyl-2-pentenoic Acid
Mængde 100 mg
InChI formel InChI=1S/C8H14O2/c1-3-5-7(6-4-2)8(9)10/h5H,3-4,6H2,1-2H3,(H,9,10)/b7-5+
Formel vægt 142.0994