Få mere at vide
(E)-N-Methylcinnamylamine-d3, TRC
Artikelnummer. 30476430
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30476430 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30476430 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2946222.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (E)-N-Methylcinnamylamine-d3
CAS 1795142-11-8
Molekylær formel C10H11D3ClN
Mængde 2.5 mg
Synonym (2E)-N-(Methyl-d3)-3-phenyl-2-propen-1-amine, (E)-N-(Methyl-d3)-3-phenyl-2-propen-1-amine
InChI formel InChI=1S/C10H13N.ClH/c1-11-9-5-8-10-6-3-2-4-7-10;/h2-8,11H,9H2,1H3;1H/b8-5+;/i1D3;
SMIL Cl.[2H]C([2H])([2H])NC\C=C\c1ccccc1
IUPAC navn (E)-3-phenyl-N-(trideuteriomethyl)prop-2-en-1-amine;hydrochloride
Molekylvægt (g/mol) 186.7
Formel vægt 186.1003