Learn More
(E)-Hexadec-9-enoic acid, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 10030-73-6 |
| Molecular Formula | C16H30O2 |
| MDL Number | MFCD00067230 |
| Quantity | 100 mg |
| SMILES | CCCCCC/C=C/CCCCCCCC(O)=O |
| Molecular Weight (g/mol) | 254.41 |
| Percent Purity | 98% |
| Chemical Name or Material | (E)-Hexadec-9-enoic acid |