Få mere at vide
(E,E)-Farnesol 11,11,11,12,12,12-d6, TRC
Artikelnummer. 30408073
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30408073 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30408073 Leverandør Toronto Research Chemicals Leverandørnr. TRCF1024325MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (E,E)-Farnesol 11,11,11,12,12,12-d6
CAS 166447-71-8
Molekylær formel C15H20D6O
Mængde 5 mg
Synonym (2E,6E)-3,7-Dimethyl-11-(methyl-d3)-2,6,10-dodecatrien-12,12,12-d3-1-ol, (E,E)-3,7-Dimethyl-11-(methyl-d3)-2,6,10-dodecatrien-12,12,12-d3-1-ol
InChI formel InChI=1S/C15H26O/c1-13(2)7-5-8-14(3)9-6-10-15(4)11-12-16/h7,9,11,16H,5-6,8,10,12H2,1-4H3/b14-9+,15-11+/i1D3,2D3
SMIL OC/C=C(C)/CC/C=C(C)/CC/C=C(C([2H])([2H])[2H])\C([2H])([2H])[2H]
Molekylvægt (g/mol) 228.4