Få mere at vide
(E)-Dimethylvinphos 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (E)-Dimethylvinphos |
| CAS | 71363-52-5 |
| Analyt- eller komponentnavne | (E)-Dimethylvinphos |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C10 H10 Cl3 O4 P |
| Koncentration | 100 μg/mL |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Phosphoric acid, (1E)-2-chloro-1-(2,4-dichlorophenyl)ethenyl dimethyl ester, Phosphoric acid, 2-chloro-1-(2,4-dichlorophenyl)ethenyl dimethyl ester, (E)-, (E)-Dimethylvinphos |
| InChI formel | InChI=1S/C10H10Cl3O4P/c1-15-18(14,16-2)17-10(6-11)8-4-3-7(12)5-9(8)13/h3-6H,1-2H3/b10-6+ |
| SMIL | COP(=O)(OC)O\C(=C\Cl)\c1ccc(Cl)cc1Cl |
| Vis mere |