missing translation for 'onlineSavingsMsg'
Få mere at vide

Vilber E-Box CX5 with Blue transiluminator and EU power cord

Artikelnummer. 31703656
Skift visning
Klik for at se tilgængelige muligheder
Til brug med (applikation):
Gel Imaging
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Til brug med (applikation)
31703656 Gel Imaging
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 31703656 Leverandør Vilber Leverandørnr. PF250202

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Designed for precise imaging performance, it offers sensitive detection and and intuitive operation through a fast, user-friendly interface.

  • High-Quality Imaging: Scientific-grade German camera provides precise detection and high-resolution imaging for accurate analysis.
  • Exceptional Sensitivity: Detects very faint DNA, RNA, or protein bands, ensuring reliable results even with low-abundance samples.
  • Publication-Ready Results: Combines sensitivity and detailed resolution to produce TIFF images suitable for scientific publications.
  • Clear Band Separation: Advanced imaging technology supports accurate distinction of closely spaced gel bands for dependable interpretation.
  • Large Touchscreen Display: Includes a very large 12.6 in. Magster touchscreen that enhances navigation and offers quick access to system functions.
  • Built-in Industrial PC (Win10): Integrated computer enables fast processing and efficient control without external hardware.
  • Durable Construction: Steel and stainless-steel darkroom ensures long-lasting performance and simplifies cleaning.
  • Advanced Camera Features: Equipped with a 5 MP sensor expandable to 20 MP, passive cooling, USB3 connectivity, 16-bit imaging with 65,536 grey levels, and a motorized zoom lens.
  • Complete and Practical: Features a compact footprint for lab benches, full network connectivity, 3D Dynamic Scan, and BioVision software for image editing and analysis.
  • Ease of Use: Stand-alone operation with one-click capture, auto-exposure, illumination control, and automatic blue light shut-off after inactivity.
  • Time-Saving: Automated features and an intuitive interface reduce hands-on work and streamline experimental routines.
  • Safe and Reliable: Automatic blue light shut-off enhances safety and protects samples from unnecessary exposure.
  • Versatile Applications: Suitable for DNA, RNA, and protein gel imaging in research, teaching, and diagnostic workflows.

Tekniske data

Vise Touchscreen
Inkluderer EU Power Cord
Til brug med (applikation) Gel Imaging
Produkttype Gel Imaging System
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.