Få mere at vide
3M™ E-A-RCap™ Headband
Shop alle 3M produkter
:
Color: Blue and Yellow; Noise Reduction Rate: 23 db
Beskrivelse
Protects workers exposed to noise. Ideal disposable hearing protection for factory visitors.
- Round pod plugs that seal the entrance of ear canal, making them more wearer acceptable
- No need to handle the plug tip making them more hygienically acceptable
- Low transmission noise through the band
- Easily stored around the neck when not in use
- Optimum attenuation for many applications
- Easy to wash and clean
- Pods can be easily replaced
Tekniske data
Tekniske data
| Farve | Blue, Yellow |
| Certificeringer/compliance | CE Approved against European Standard EN352-2:1993. |
| Noise Reduction Rating (NRR) | 23 dB |
| Stikform | Rounded |
| Produkttype | Ear Plug |
| Materiale | Polycarbonate (Band) / Polyurethane Foam (Ear tip) |