Få mere at vide
(E)-5-(2-Carboxvinyl)uracil, TRC
Artikelnummer. 30368146
Skift visning
Mængde:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30368146 250 mg
30364724 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30368146 Leverandør Toronto Research Chemicals Leverandørnr. TRCC183225250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (E)-5-(2-Carboxvinyl)uracil
CAS 57412-59-6
Molekylær formel C7 H6 N2 O4
Mængde 250 mg
InChI formel InChI=1S/C7H6N2O4/c10-5(11)2-1-4-3-8-7(13)9-6(4)12/h1-3H,(H,10,11)(H2,8,9,12,13)/b2-1+
SMIL OC(=O)\C=C\C1=CNC(=O)NC1=O
IUPAC navn (E)-3-(2,4-dioxo-1H-pyrimidin-5-yl)prop-2-enoic acid
Molekylvægt (g/mol) 182.13
Formel vægt 182.0328