Få mere at vide
(E)-4-Methyl-2-phenyl-2-pentenal (Stabilized with Alpha-tocopherol), TRC
Artikelnummer. 30471496
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30471496 50 mg
30464941 100 mg
30466281 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30471496 Leverandør Toronto Research Chemicals Leverandørnr. TRCM22035350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1474032-26-2
Molekylær formel C12H14O
Molekylvægt (g/mol) 174.24
Synonym (alphaE)-alpha-(2-Methylpropylidene)benzeneacetaldehyde
IUPAC navn (E)-4-methyl-2-phenylpent-2-enal
SMIL CC(C)C=C(C=O)c1ccccc1
Tekniske data
Kemisk navn eller materiale (E)-4-Methyl-2-phenyl-2-pentenal
Mængde 50 mg
InChI formel InChI=1S/C12H14O/c1-10(2)8-12(9-13)11-6-4-3-5-7-11/h3-10H,1-2H3/b12-8-
Formel vægt 174.1045