missing translation for 'onlineSavingsMsg'
Få mere at vide

(E)-3-Hexenyl Butanoate, TRC

Artikelnummer. 30407579
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30407579 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30407579 Leverandør Toronto Research Chemicals Leverandørnr. TRCH2947881G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (E)-3-Hexenyl Butanoate
CAS 53398-84-8
Molekylær formel C10 H18 O2
Mængde 1 g
Synonym (E)-3-Hexenyl Butanoate, (E)-Butanoic Acid 2-hexenyl Ester, (E)-Hex-3-en-1-yl Butyrate, Butanoate-trans-3-hexene ester, E-3-Hexenyl Butyrate, trans-3-Hexenyl Butanoate, trans-3-Hexenyl Butyrate
InChI formel InChI=1S/C10H18O2/c1-3-5-6-7-9-12-10(11)8-4-2/h5-6H,3-4,7-9H2,1-2H3/b6-5+
SMIL CCCC(=O)OCC\C=C\CC
IUPAC navn [(E)-hex-3-enyl] butanoate
Molekylvægt (g/mol) 170.249
Formel vægt 170.131
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.