Få mere at vide
(E)-2-[2-(Bromomethyl)phenyl]-2-(methoxyimino)acetic Acid Methyl Ester, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (E)-2-[2-(Bromomethyl)phenyl]-2-(methoxyimino)acetic Acid Methyl Ester |
| CAS | 133409-72-0 |
| Molekylær formel | C11H12BrNO3 |
| Mængde | 250 mg |
| Synonym | (E)-Methyl 2-(2-Bromomethylphenyl)-2-methoximinoacetate, (E)-Methyl 2-(2-Bromomethylphenyl)glyoxylate O-Methyloxime, (E)-Methyl 2-Bromomethyl-alpha-methoxyiminophenylacetate, (E)-2-(Bromomethyl)phenylglyoxylic Acid Methyl Ester O-methyloxime, (E)-2-Methoxyimino-2-[2-(bromomethyl)phenyl]acetic Acid Methyl Ester |
| InChI formel | InChI=1S/C11H12BrNO3/c1-15-11(14)10(13-16-2)9-6-4-3-5-8(9)7-12/h3-6H,7H2,1-2H3 |
| SMIL | CO\N=C(\C(=O)OC)/c1ccccc1CBr |
| IUPAC navn | methyl 2-[2-(bromomethyl)phenyl]-2-methoxyiminoacetate |
| Molekylvægt (g/mol) | 286.12 |
| Formel vægt | 285.0001 |