missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Dynapyrazole A

Description
Dynapyrazole A is a dynein inhibitor. Inhibits dynein 1 and dynein 2-dependent microtubule gliding (IC50 values are 2.3 and 2.6 μM, respectively). Inhibits hedgehog signaling in vitro (IC50 = 1.9 μM). Displays reversible inhibition of retrograde and anterograde intraflagellar transport in Chlamydomonas; displays stronger inhibition of retrograde transport.
Specifications
Specifications
| Kemisk navn eller materiale | 2-[1-(4-Chlorophenyl)cyclopropyl]-4,5-dihydro-7-iodo-5-oxopyrazolo[1,5-a]quinazoline-3-carbonitrile |
| CAS | 2226517-75-3 |
| Mængde | 25 mg |
| Mål | Hedgehog Signaling Inhibitors |
| Molekylær formel | C20H12ClIN4O |
| Formel vægt | 3333 |
| Renhed | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?