Learn More
ABEBA™ Dynamic White Occupational Clogs Ob ESD
White dynamic occupational clogs with non-foldable heel strap, replaceable active comfort insole, slip resistant TPU outsole
528.00 DKK
Specifications
Color | White |
---|---|
Material | Knitted Fabric |
Certifications/Compliance (Europe) | ESD dissipative to EN 61340 Climate class 1, Comply to EN ISO 20347: 2012 OB SRC |
Gender | Unisex |
Ankle Style | Clogs |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
17111365
|
ABEBA™
37329/35 |
35 |
528.00 DKK
1 pair |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||||
17121365
|
ABEBA™
37329/36 |
36 |
528.00 DKK
1 pair |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||||
17111375
|
ABEBA™
37329/45 |
45 |
528.00 DKK
1 pair |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||||
17121375
|
ABEBA™
37329/46 |
46 |
528.00 DKK
1 pair |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||||
17131375
|
ABEBA™
37329/47 |
47 |
528.00 DKK
1 pair |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||||
17141375
|
ABEBA™
37329/48 |
48 |
528.00 DKK
1 pair |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||||
17131365
|
ABEBA™
37329/37 |
37 |
528.00 DKK
1 pair |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||||
17141365
|
ABEBA™
37329/38 |
38 |
528.00 DKK
1 pair |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||||
17151365
|
ABEBA™
37329/39 |
39 |
528.00 DKK
1 pair |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||||
17161365
|
ABEBA™
37329/40 |
40 |
528.00 DKK
1 pair |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||||
17171365
|
ABEBA™
37329/41 |
41 |
528.00 DKK
1 pair |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||||
17181365
|
ABEBA™
37329/42 |
42 |
528.00 DKK
1 pair |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||||
17191365
|
ABEBA™
37329/43 |
43 |
528.00 DKK
1 pair |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||||
17101375
|
ABEBA™
37329/44 |
44 |
528.00 DKK
1 pair |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||||
Description
- Breathable, lightweight, comfortable occupational clogs
- Comply to EN ISO 20347: 2012, OB SRC
- ESD dissipative according to DIN EN 61340
- Slip resistant TPU outsole
- Good cushioning
- Energy absorption in heel area
- Water repellent white knitted textile
- Non foldable heel strap, adjustable
- Removable active comfort insole
Specifications
White | |
ESD dissipative to EN 61340 Climate class 1, Comply to EN ISO 20347: 2012 OB SRC | |
Clogs | |
OB SRC ESD |
Knitted Fabric | |
Unisex | |
Water Repellent |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
ABEBA™ Dynamic White Occupational Clogs Ob ESD