Learn More
Abeba™ Dynamic 7310 Shoes
Special respiration absorbing lining allows the foot to breathe. Includes TPU sole with high slip resistance.
Brand: Abeba™ 7310/43
This item is not returnable.
View return policy
Description
- CE EN ISO 20347:2012 OB A, E, FO SRC
- Smooth leather white
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3574)
- Sanitized treated
- Antistatic
- Heel strap foldable, adjustable
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC | |
Leather | |
Slip-resistant | |
7310 |
White | |
43 | |
Polyurethane |
Product Content Correction
Product Title
Abeba™ Dynamic 7310 Shoes > Size: 43
Spot an opportunity for improvement?Share a Content Correction