missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Dynabeads™ MAX E. coli O157 Kit

Dynabeads™ MAX E. coli O157 Kit

Brand:  Applied Biosystems™ A10715

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10444995

  • 5088.05 DKK / Each


Estimated Shipment
11-03-2021
Add to basket

Description

Description

Dynabeads™ MAX E.Coli O157 provides a powerful sample preparation method to improve PCR detection and quantification of E.Coli O157:H7 in food, feces, water and other environmental samples.
Based on well known and widely used immunomagnetic separation (IMS) technology, Dynabeads™ MAX E.Coli O157 provides a selective E.Coli O157:H7 capture step that delivers significant improvements in PCR analytical process time, as well as the sensitivity and consistency of results. With Dynabeads™ MAX E.Coli O157, you'll:

  • Perform high throughput, rapid, and effective E.Coli O157:H7 sample preparation
  • Significantly shorten sample pre-enrichment time
  • Improve detection sensitivity in PCR sample analysis
  • Significantly reduce overall analysis time
  • Can be carried out manually or automated using the BeadRetriever™ System
  • Specifications

    Specifications

    Polyclonal
    Bacteria (Intact)
    Immunomagnetic Separation
    Small Volume Microbial Samples
    1-3 mL
    E. coli O157:H7
    Dynabeads™ MPC™, KingFisher mL Food Protection Purification System, DynaMag™-2
    Environmental Sample, Feces, Food, Water Samples
    Each unit of product contains 5 vials X 1 ml Dynabeads™ MAX anti-E. coli O157 and 80 ml 10X Sample Buffer.

    Store at 2–8°C.
    Not High-throughput Compatible (Manual), Automated Protocols
    250 Tests
    DYNAL™, Dynabeads™
    Room Temperature
    E. coli
    5mL
    Videos
    SDS
    Documents

    For Research Use Only. Not for use in diagnostic procedures.