missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ Intact Virus Enrichment

Dynabeads Intact Virus Enrichment beads are 1-μm strong anion exchange magnetic beads that bind negatively charged vesicles and molecules.

6160.00 DKK - 21830.00 DKK

Specifications

Concentration 40 mg/mL
Description Strong anion exchange magnetic beads
Material Magnetic Polystyrene
Content And Storage Dynabeads Intact Virus Enrichment Beads are supplied in 30 mM NaCl with 0.05% NaN3. Store at 2°C to 8°C.

Diameter (Metric) 1 μm
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17176088
View Documents Promotion Details
Invitrogen™
10700D
100 reactions
6160.00 DKK
100 reactions
Please to purchase this item. Need a web account? Register with us today!
17186088
View Documents
Invitrogen™
10701D
500 reactions
21830.00 DKK
500 reactions
Please to purchase this item. Need a web account? Register with us today!
Description
Show More
Specifications

Specifications

40 mg/mL
Magnetic Polystyrene
1 μm
High-throughput Compatible
Virus Enrichment
Dynabeads Intact Virus Enrichment
For Research Use Only
Strong anion exchange magnetic beads
Dynabeads Intact Virus Enrichment Beads are supplied in 30 mM NaCl with 0.05% NaN3. Store at 2°C to 8°C.

KingFisher Sample Purification Systems, DynaMag Magnets
Strong positively charged surface
Negatively charged viruses, exosomes, proteins
ISO9001 and ISO13485
Monosized 1 μm (CV <5%)
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 24 months from date of manufacture

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Dynabeads™ Intact Virus Enrichment

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.