Få mere at vide
Dutasteride Beta-Dimer, TRC
Artikelnummer. 30135819
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135819 5 mg
30132387 1 mg
30004336 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135819 Leverandør Toronto Research Chemicals Leverandørnr. TRCD7350205MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1648593-70-7
Molekylær formel C46 H55 F6 N3 O4
Molekylvægt (g/mol) 827.94
Synonym 1H-Indeno[5,4-f]quinoline-7-carboxamide, N-[2,5-bis(trifluoromethyl)phenyl]-2,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-tetradecahydro-4a,6a-dimethyl-2-oxo-1-[[(4aR,4bS,6aS,7S,9aS,9bS,11aR)-2,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-tetradecahydro-4a,6a-dimethyl-2-oxo-1H-indeno[5,4-f]quinolin-7-yl]carbonyl]-, (4aR,4bS,6aS,7S,9aS,9bS,11aR)-, N-[2,5-Bis(trifluoromethyl))phenyl]-3-oxo-4-[3-oxo-4-aza-5α-androst-1-ene-17β-carbonyl]-4-aza-5α-androst-1-ene-17β-carboxamide, Dutasteride Dimer 1, Dutasteride Imp. I (EP), Dutasteride β-Dimer
IUPAC navn (1S,3aS,3bS,5aR,9aR,9bS,11aS)-6-[(1S,3aS,3bS,5aR,9aR,9bS,11aS)-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carbonyl]-N-[2,5-bis(trifluoromethyl)phenyl]-9a,11a-dimethyl-7-oxo-2,3,3a,3b,4,5,5a,9b,10,11-decahydro-1H-indeno[5,4-f]quinoline-1-carboxamide
SMIL C[C@]12CC[C@H]3[C@@H](CC[C@H]4N(C(=O)[C@H]5CC[C@H]6[C@@H]7CC[C@H]8NC(=O)C=C[C@]8(C)[C@H]7CC[C@]56C)C(=O)C=C[C@]34C)[C@@H]1CC[C@@H]2C(=O)Nc9cc(ccc9C(F)(F)F)C(F)(F)F
Tekniske data
Kemisk navn eller materiale Dutasteride beta-dimer
Mængde 5 mg
InChI formel InChI=1S/C46H55F6N3O4/c1-41-19-16-30-26(27(41)9-11-32(41)39(58)53-34-23-24(45(47,48)49)5-8-31(34)46(50,51)52)7-14-36-44(30,4)22-18-38(57)55(36)40(59)33-12-10-28-25-6-13-35-43(3,21-17-37(56)54-35)29(25)15-20-42(28,33)2/h5,8,17-18,21-23,25-30,32-33,35-36H,6-7,9-16,19-20H2,1-4H3,(H,53,58)(H,54,56)/t25-,26-,27-,28-,29-,30-,32+,33+,35+,36+,41-,42-,43+,44+/m0/s1
Formel vægt 827.4097