Få mere at vide
Dutasteride-13C6, TRC
Artikelnummer. 30370869
Skift visning
Mængde:
0.25 mg
1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30370869 0.25 mg
30370270 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30370869 Leverandør Toronto Research Chemicals Leverandørnr. TRCD7350020.25MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1217685-27-2
Molekylær formel C2113C6H30F6N2O2
Molekylvægt (g/mol) 534.49
Synonym (4aR,4bS,6aS,7S,9aS,9bS,11aR)-N-[2,5-Bis(trifluoromethyl)phenyl]-2,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-tetradecahydro-4a,6a-dimethyl-2-oxo-1H-indeno[5,4-f]quinoline-7-carboxamide-13C6, (5α,17β)-N-[2,5-Bis(trifluoromethyl)phenyl]-3-oxo-4-azaandrost -1-ene-17-carboxamide-13C6, Avodart-13C6, Avolve-13C6, GG 745-13C6, GI 198745-13C6, Veltride-13C6
SMIL [H][C@@]12CC[C@@]3([H])NC(C=C[C@]3(C)[C@@]1([H])CC[C@@]4(C)[C@@]2([H])CC[C@@H]4C(N[13C]5=[13CH][13C](C(F)(F)F)=[13CH][13CH]=[13C]5C(F)(F)F)=O)=O
Tekniske data
Kemisk navn eller materiale Dutasteride-13C6
Mængde 0.25 mg
InChI formel InChI=1S/C27H30F6N2O2/c1-24-11-9-17-15(4-8-21-25(17,2)12-10-22(36)35-21)16(24)6-7-19(24)23(37)34-20-13-14(26(28,29)30)3-5-18(20)27(31,32)33/h3,5,10,12-13,15-17,19,21H,4,6-9,11H2,1-2H3,(H,34,37)(H,35,36)/t15-,16-,17-,19+,21+,24-,25+/m0/s1/i3+1,5+1,13+1,14+1,18+1,20+1