Learn More
DWK Life Sciences DURAN™ Vacuum Receiver Adapter, straight, with 2 standard ground joints, and screw-on plastic hose connection
The receiver adapter is suitable for transferring the distillate from the condenser to the receiving flask.
Brand: DWK Life Sciences 241312106
382.85 DKK valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.04300kg
Description
Part of a distillation assembly is, among other things, the receiver adapter, which is connected downstream at the end of the condenser and guides the distillate into the receiving flask. If vacuum distillation is performed, the vacuum pump is ideally connected to the receiver adapter.
- Tight closure due to standard ground joint
- Straight
- With two standard ground joint and screw-on plastic hose connection
- Fired-on, highly durable white print
Specifications
Autoclavable | |
8.6 mm | |
1 Ea. |
Distillation | |
Borosilicate Glass (3.3), PBT, Polypropylene, Silicone | |
NS 14/23 |