Learn More
DWK Life Sciences DURAN™ Two Way Stopcock, double oblique bore, ground, without key
Ground 2-way valves without plug.
Brand: DWK Life Sciences 286425204
Description
Ground DURAN™ patent valves are 2-way valves, which are frequently used for the construction of steering lines (vacuum inert gas lines, nitrogen distribution rakes). We recommend the use of PRODURAN™ valves for particularly high leakage rate requirements.
- Valve sleeve ground, without plug
- 2-way valve
- Glass articles for further processing
Specifications
4 mm | |
18.8 NS | |
Suitable for installation in glass apparatus for vacuum applications, e.g. for the construction of steering lines (vacuum inert gas lines, nitrogen distribution rakes) |
10 mm | |
1.5 mm |