Learn More
DWK Life Sciences DURAN™ Two Way Stopcock, double oblique bore, ground, without key
Ground 2-way valves without plug.
Brand:  DWK Life Sciences 286425204
 View more versions of this product
Product Code. 15884728
  • 2490.00 DKK / Pack of 10
Only null left
Estimated Shipment: 30-06-2025
Add to basket
This item is not returnable. View return policy
Description
Description
Ground DURAN™ patent valves are 2-way valves, which are frequently used for the construction of steering lines (vacuum inert gas lines, nitrogen distribution rakes). We recommend the use of PRODURAN™ valves for particularly high leakage rate requirements.
  • Valve sleeve ground, without plug
  • 2-way valve
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4 mm
18.8 NS
Suitable for installation in glass apparatus for vacuum applications, e.g. for the construction of steering lines (vacuum inert gas lines, nitrogen distribution rakes)
10 mm
1.5 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences DURAN™ Two Way Stopcock, double oblique bore, ground, without key > Double oblique bore 4 mm, NS 18.8