Learn More
DWK Life Sciences DURAN™ Three-way Stopcock, complete with SBW-J key, T-bore
3-way tap with threaded plug.
Brand: DWK Life Sciences 286416405
Description
T-valves are the most common form of 3-way valves. The DURAN™ three-way valve offers the following adjustment options: 1. Connection of two different attachments 2. Connection of all 3 attachments 3. Close all 3 attachments
- 3-way valve with threaded plug
- SBW-J glass threaded tap plug with T-bore
- Three-way valve offers the following setting options: 1. Connection of two different attachments 2. Connection of all 3 attachments 3. Close all 3 attachments
- Glass articles for further processing
Specifications
2.0 mm | |
Borosilicate Glass (3.3), Polypropylene | |
14.5 NS | |
5/Pk. |
8 mm | |
Not Autoclavable | |
1.5 mm | |
Installation in glass apparatus and devices |