Learn More
DWK Life Sciences DURAN™ Three-way Stopcock, complete with SBW-J key, T-bore
3-way tap with threaded plug.
Brand:  DWK Life Sciences 286416405
 View more versions of this product
Product Code. 15814728
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
T-valves are the most common form of 3-way valves. The DURAN™ three-way valve offers the following adjustment options: 1. Connection of two different attachments 2. Connection of all 3 attachments 3. Close all 3 attachments
  • 3-way valve with threaded plug
  • SBW-J glass threaded tap plug with T-bore
  • Three-way valve offers the following setting options: 1. Connection of two different attachments 2. Connection of all 3 attachments 3. Close all 3 attachments
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2.0 mm
Borosilicate Glass (3.3), Polypropylene
14.5 NS
5/Pk.
8 mm
Not Autoclavable
1.5 mm
Installation in glass apparatus and devices
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences DURAN™ Three-way Stopcock, complete with SBW-J key, T-bore > T-bore 2 mm