Få mere at vide
DWK Life Sciences DURAN™ Standard Socket, double, finished
Shop alle DWK Life Sciences produkter
Ledstørrelse:
18 mm
22 mm
Pakningsstørrelse:
Pakke med 50
Beskrivelse
The DURAN™ double cores, ground, are an old, less common type of design according to DIN 12249 Form C. It primarily serves the purpose of saving material.
- According to DIN 12249 Form C
- Glass article with two sleeves enables material savings
- Very low leakage rate
- Minimized grease requirement
- Tight closure due to standard ground joint
- Glass articles for further processing
Tekniske data
Tekniske data
| Mængde | 50/Pk. |
| Varebeskrivelse | Installation in glass apparatus and devices |
| Ledstørrelse | 18 mm |
| Tykkelse (metrisk) væg | 1.8 mm |