Få mere at vide
DWK Life Sciences DURAN™ Standard kegle med spids, færdig
Artikelnummer. 10182121 Shop alle DWK Life Sciences produkter
Ledstørrelse:
6 mm
8 mm
11 mm
13 mm
17 mm
22 mm
26 mm
30 mm
36 mm
40 mm
46 mm
50 mm
56 mm
65 mm
80 mm
90 mm
Pakningsstørrelse:
Hver
Pakke med 10
Pakke med 50
Artikelnummer. 10182121
Brand: DWK Life Sciences 286166004
Kerneskåret med forlængelse.
The DURAN™ cores with outlet comply with DIN 12249 Form G. The process has the function of a drip ring. It prevents the liquid from running down the inner wall of the vessel. For this purpose, the outlet is usually cut at an angle and fused. At the same time, mediums do not come into contact with the greased ground surface. The additional outlet can also be used as an attachment point for extensions for various purposes, e.g. as an inlet pipe.
  • Ground (finer grinding quality, less roughness depth)
  • The additional drain on the grinding enables a clean drain/leakage of liquids without wetting the grinding.
  • Tight closure due to standard ground joint
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Tekniske data
Mængde 50/Pk.
Varebeskrivelse The additional outlet can also be used as an attachment point for extensions for various purposes, e.g. as an inlet pipe; Installation in glass apparatus and devices
Ledstørrelse 22 mm
Tykkelse (metrisk) væg 1.8 mm