Learn More
DWK Life Sciences DURAN™ Standard Cone, double, finished
For a rigid, self-supporting connection.
Brand:  DWK Life Sciences 286173205
2807.25 DKK valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 5.00000kg
 View more versions of this product
Product Code. 15854598
  • 2955.00 DKK / Pack of 50
Estimated Shipment: 21-05-2024
to see stock.
Add to basket
Description
Description
The DURAN™ double cores, ground, are an old, less common type of design according to DIN 12249 Form D. Their primary purpose is to save material.
  • According to DIN 12249 Form D
  • Ground (finer grinding quality, less roughness depth)
  • Glass article with two cores enables material savings
  • Tight closure due to standard ground joint
  • Glass articles for further processing
Specifications
Specifications
50/Pk.
Installation in glass apparatus and devices
17 mm
1.8 mm