Learn More
DWK Life Sciences DURAN™ Standard Cone, diamond-ground
Designed for a rigid, self-supporting connection with lower requirements.
Brand:  DWK Life Sciences 286156303
1349.00 DKK valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15854568
  • 1420.00 DKK / Pack of 50
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
Description
Description
The conical ground joint of sleeve and core is a rigid, self-supporting joint. DURAN™ DG cores are not ground according to DIN 12249 and represent the cost-effective alternative to DURAN™ cores. Ground joints with ground grease or PTFE sleeves should always be used. KECK™ Conical ground joint clamps are the perfect supplement for securing the conical ground joint.
  • Diamond cut (coarser cut quality, more roughness depth)
  • Cost-effective alternative to DURAN™ cores (not according to DIN)
  • Tight closure due to standard ground joint
  • Glass articles for further processing
Specifications
Specifications
50/Pk.
Installation in glass apparatus and devices
22 mm
1.8 mm