Learn More
DWK Life Sciences DURAN™ Single way Stopcock, ground and polished, without key
Ground and polished shut-off taps without plug for grease free operation.
1130.00 DKK - 6380.00 DKK
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
Autoclavable | Yes |
For Use With (Application) | Coarse dosing of liquids; Installation in glass apparatus and devices |
Product Code | Brand | Closure Size | Plug Bore | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Size | Plug Bore | Price | Quantity & Availability | ||||
15804768
|
DWK Life Sciences
286440405 |
18.8 NS | 4 mm |
3420.00 DKK
Pack of 25 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
15884758
|
DWK Life Sciences
286439807 |
14.5 NS | 4 mm |
6380.00 DKK
Pack of 50 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
15814768
|
DWK Life Sciences
286440602 |
18.8 NS | 6 mm |
3765.00 DKK
Pack of 25 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
15844758
|
DWK Life Sciences
286438005 |
10 NS | 1.5 mm |
1130.00 DKK
Pack of 10 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
15854758
|
DWK Life Sciences
286439001 |
12.5 NS | 2 mm |
5410.00 DKK
Pack of 50 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
15824768
|
DWK Life Sciences
286441007 |
21.5 NS | 6 mm |
1245.00 DKK
Pack of 5 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
15844768
|
DWK Life Sciences
286441607 |
29.2 NS | 10 mm |
2055.00 DKK
Pack of 5 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
15834768
|
DWK Life Sciences
286441401 |
24 NS | 8 mm |
1615.00 DKK
Pack of 5 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
15874758
|
DWK Life Sciences
286439601 |
14.5 NS | 2.5 mm |
5880.00 DKK
Pack of 50 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
Description
Ground and polished DURAN™ single way stopcocks are shut-off taps which are combined with PTFE valve plugs to allow grease free operation.
- Ground shut-off taps without tap plug
- Fat-free working through the polished tap surface
- Taper 1:10
- Glass articles for further processing
Specifications
Borosilicate Glass (3.3) | |
Yes | |
Coarse dosing of liquids; Installation in glass apparatus and devices |