Learn More
DWK Life Sciences DURAN™ Separating Funnel Stopcock with short inlet, complete with PTFE key
For easier attachment to DURAN™ separating funnel raw bodies.
5995.00 DKK - 12355.00 DKK
Specifications
Material | Borosilicate Glass (3.3), PTFE |
---|---|
Autoclavable | Autoclavable |
No. per Pack | 25/Pk. |
For Use With (Application) | For attachment to DURAN™ separating funnel raw bodies |
Product Code | Brand | Closure Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Size | Price | Quantity & Availability | ||||||
15874748
|
DWK Life Sciences
286432602 |
12.5 NS | N/A | |||||||
15884748
|
DWK Life Sciences
286432808 |
14.5 NS |
6830.00 DKK
Pack of 25 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
15894748
|
DWK Life Sciences
286433007 |
18.8 NS |
10700.00 DKK
Pack of 25 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
15804758
|
DWK Life Sciences
286433204 |
21.5 NS |
12355.00 DKK
Pack of 25 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
Description
DURAN™ Separating funnel taps with short funnel are components for the production of Gilson separating funnels with a particularly strong approach to reducing the risk of breakage with a conical shape right down to the socket of the valve. In the polished version with PTFE plug suitable for grease free working.
- Separating funnel tap sleeve with PTFE plug
- with short approach funnel
- With a particularly strong approach to reducing the risk of breakage
- Glass articles for further processing
Specifications
Borosilicate Glass (3.3), PTFE | |
Autoclavable | |
25/Pk. | |
For attachment to DURAN™ separating funnel raw bodies |