Learn More
DWK Life Sciences DURAN™ NMR Tubes, for four application frequencies, with closures
Product Code. 15271819 Shop All DWK Life Sciences Products
Change view
Click to view available options
Field Strength:
300 MHz
400 MHz
500 MHz
Unit Size:
Pack of 250
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Field Strength Unit Size
15271819 400 MHz Pack of 250
11361475 300 MHz Pack of 250
11880970 500 MHz Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15271819 Supplier DWK Life Sciences Supplier No. 231700211
  • ONLINE EXCLUSIVE PRICE
    19610.00 DKK
    17240.17 DKK / Pakke med 250
    Spare 2369.83 DKK
    12% Slukket
View all promotions
Forventet forsendelse: 07-12-2026
NMR tubes, the sample or solution vessels for resonance frequency measurement, are extremely precise and are subject to narrow tolerances for straightness, wall thickness and wall thickness distribution.
Nuclear magnetic resonance spectroscopy (NMR-spectroscopy) is a spectroscopic method to elucidate the structure of molecules and compounds. Due to electronic interactions of single atoms with the neighbouring atoms, structural conclusions can be drawn. In addition to structure elucidation, the concentration and dynamics of molecules can be determined with this method. This analytical method requires very precise tubes with a constant diameter, a uniform wall thickness and excellent straightness across the entire tube.
  • Close tolerances and accuracy, especially to their straightness, wall thickness and wall thickness distribution
  • Supplied with cap
Specifications
Material Borosilicate Glass (3.3), EVA
Product Type NMR Tube
Diameter (Metric) Outer 4.95±0.05 mm
Field Strength 400 MHz
For Use With (Application) Nuclear magnetic resonance spectroscopy (NMR spectroscopy); Storage and transport of measurement solutions; Designed for use in low to most mid field NMR spectrometers
Quantity 250/Pk.
Model Professional with Retrace Code