Få mere at vide
Witeg™ Duran Glass Separatory Funnels
Artikelnummer. 10081122
Skift visning
Kapacitet (metrisk):
50 ml
100 ml
250 ml
1000 ml
2000 ml
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Kapacitet (metrisk)
10081122 250 ml
10750415 100 ml
10632563 1000 ml
10421763 50 ml
10787534 2000 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 10081122 Leverandør Witeg™ Leverandørnr. 2 677 025
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Witeg™ Duran™ Glass Separatory Funnels are used in liquid-liquid extractions to separate (partition) the components of a mixture into two immiscible solvent phases of different densities.
  • Conical shape
  • Graduated
  • Economy-needle valve stopcock
  • PTFE-needle valve
  • Screw-thread and cap
Tekniske data
Kapacitet (metrisk) 250 ml
Materiale Duran Glass, Borosilicate Glass
Mængde 1Pack
Produkttype Separatory Funnel