Få mere at vide
DWK Life Sciences DURAN™ Udvidelsesadapter, fatning/kegle
Shop alle DWK Life Sciences produkter
Ledstørrelse:
Fatning NS 14/23, Kegle NS 19/26
Fatning NS 14/23, Kegle NS 24/29
Fatning NS 19/26, Kegle NS 24/29
Fatning NS 19/26, Kegle NS 29/32
Fatning NS 24/29, Konus NS 29/32
Fatning NS 29/32, Kegle NS 34/35
Fatning NS 29/32, Kegle NS 45/40
Fatning NS 29/32, Kegle NS 50/42
Fatning NS 29/32, Kegle NS 60/46
Fatning NS 29/32, Kegle NS 71/51
Muffe NS 14/23, Konus NS 29/32
Topnøgle NS 24/29, Konus NS 45/40
Pakningsstørrelse:
Hver
Pakke med 10
Beskrivelse
Adapters are used to connect two components with different NS sizes. DURAN™ expansion pieces allow the connection of a smaller sleeve to a larger core.
- Expansion pieces allow the connection of a smaller sleeve to a larger core and thus the use of differently sized ground joint components.
Tekniske data
Tekniske data
| Mængde | 10/Pk. |
| Varebeskrivelse | Connection of two components with different standard ground joint sizes |
| Ledstørrelse | Muffe NS 14/23, Konus NS 29/32 |