Learn More
Hirschmann™ DURAN™ Class AS Micro Burettes Blue Graduations
Excellent for dispensing measured amounts of chemical solutions in the laboratory. Manufactured from durable DURAN™ glass tubing.
Brand: Hirschmann™ 3310152
Additional Details : Weight : 0.25000kg
Description
- Schellbach
- Bang pattern
- Main point ring graduation
- With blue graduation
- Acc. to DIN EN ISO 385
- Support base
- Conformity certified, with dated batch identification
Specifications
Micro Burettes acc. to Bang, Class AS, conformity certified | |
DURAN™, burette and stopcock key | |
Bang, Main Point Ring Graduation, Schellbach Stripes, Socket | |
Blue | |
Glass | |
Class AS |
5 mL | |
0.02 mL | |
Lateral stopcock, standard ground glass key | |
Lateral | |
±0.01 mL | |
DIN EN ISO 385 |