missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Burette Reservoir Bottle

Catalog Number p-7189468
532.00 DKK valid until 2024-12-31
Use promo code "23258" to get your promotional price.


Click to view available options
Color:
Amber
Clear
This item is not returnable. View return policy
This item is not returnable. View return policy

The Burette reservoir bottle serves as a storage vessel for solutions during titration using an automatic burette.

The original DURAN™ burettes are made of highly chemically resistant borosilicate glass 3.3. Their precise scaling makes them particularly suitable for titrimetric purposes. The amount of liquid required for titration, but previously unknown, can be read off very accurately after the reaction has been completed. Adjustment is made to drain (Ex) for + 20°C reference temperature. This means that the quantities can be taken exactly according to the scale, since the adhesion of the liquid to the glass was taken into account during calibration. The tolerances for the volume correspond to the error limits of ISO and DIN. DURAN™ burettes are available with glass or PTFE keys. Unlike glass keys, PTFE keys do not have to be greased, which simplifies work in the laboratory.

  • Replacement bottle for automatic burettes
  • Supplied without accessories
Diameter (Metric) 160 mm
Capacity (Metric) 2000 mL
Color Clear
For Use With (Application) Titration; Volumetric measurement; Measurement analysis
Includes With standard ground joint 29/32, clear
Material Borosilicate Glass (3.3)
Quantity 1 Ea.
Autoclavable Autoclavable
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'