Learn More
Lenz Laborglasinstrumente™ Adapters, Bent
Provides for quick and easy leak-tight connections to apparatus in the laboratory. Lenz Laborglasinstrumente™ Adapters, Bent are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 05095004
Additional Details : Weight : 0.02700kg
Product Code. 10693691
  • 122.96 DKK / Each
Estimated Shipment: 16-05-2024
to see stock.
Add to basket
Description
Description
  • With NS cone and bent glass olive at a 90° angle
  • Corrosion-free and resistant to thermal shock
  • With conical joint, featuring high taper accuracy and excellent tightness
Specifications
Specifications
Adapter
Duran
With NS cone and bent glass olive (angle 90°)