Learn More
DuraCal™ 4.01 Value pH Buffer, Hamilton™
Economical choice since only about 20mL of buffer is used per calibration. Hamilton™ DuraCal™ 4.01 Value pH Buffers have actual values determined by a DKD laboratory, accredited for pH measurement.
Brand:  Hamilton 238917
Quantity 3 x 500 mL
Product Code. 10714673
  • 1200.00 DKK / Pack of 3
Edge
Description
Description
  • 4.01 pH value
  • Immune to micro-organisms
  • Expiration date on the bottle
  • First class certificate with traceability to international standards
Specifications
Specifications
4.01
3 x 500 mL
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
DuraCal™ 4.01 Value pH Buffer, Hamilton™ > Quantity: 3 x 500mL