missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Tocris Bioscience™ dTAGV-1

Beskrivelse
dTAGV-1 is a degrader targeting mutant FKBP12F36V fusion proteins. Comprises a ligand selective for F36V single-point mutated FKBP12, a linker and a von Hippel-Lindau (VHL)-binding ligand. Induces potent and selective degradation of FKBP12F36V fusion proteins in vitro and in vivo. Selectively degrades FKBP12F36V-EWS/FLI fusion proteins and inhibits cell proliferation in FKBP12F36V-EWS/FLI-expressing Ewing sarcoma cells. Hydrochloride salt (Cat.No. 7374) available; suitable for in vivo use. Negative control dTAGV-1-NEG (Cat. No. 6915) also available. FKBP12F36V can be expressed as a fusion with a target protein of interest using genome engineering techniques, via transgene expression or CRISPR-mediated locus-specific knock-in. Custom knock-in cell lines for the dTAG and aTAG platforms are available from our sister brand R&D Systems . Email TPD@bio-techne.com to enquire. Plasmid vectors for the lentiviral expression and CRISPR-mediated knock-in of FKBP12F36V are available from Addgene.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (R)-3-(3,4-Dimethoxyphenyl)-1-(2-(2-((7-((-1-((2 S,4 R)-4-hydroxy-2-((-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-7-oxoheptyl)amino)-2-oxoethoxy)phenyl)propyl -1-(-2-(3,4,5-trimethoxyph) |
| Mængde | 5 mg |
| Mål | TAG Degradation Platforms |
| Molekylær formel | C68H90N6O14S CF3CO2H |
| Renhed | 0.98 |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?