missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
dsRNA Antibody (J5), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11396R
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
dsRNA Monoclonal antibody specifically detects dsRNA in Broad Species Reactivity samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunoblot
Tekniske data
| dsRNA | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Broad Species Reactivity | |
| Purified |
| ELISA, Immunohistochemistry, Immunofluorescence, Immunoblot | |
| J5 | |
| ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoblotting | |
| Double-stranded RNA | |
| Female DBA/2 mice were injected intraperitonially with a mixture of 50 ug L-dsRNA and 75 ug methylated bovine serum albumin, emulsified in complete Freund's adjuvant. After several boosts spleen cells were fused with Sp2/0-Agl4 myeloma cells to generate the hybridoma clone. | |
| 0.2 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2b κ |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion