Få mere at vide
DSRM-3716, TRC
Artikelnummer. 30011075
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011075 50 mg
30007196 100 mg
30003714 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30011075 Leverandør Toronto Research Chemicals Leverandørnr. TRCI70732050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 58142-99-7
Molekylær formel C9 H6 N I
Molekylvægt (g/mol) 255.055
Synonym DSRM-3716
IUPAC navn 5-iodoisoquinoline
SMIL Ic1cccc2cnccc12
Tekniske data
Kemisk navn eller materiale 5-Iodoisoquinoline
Mængde 50 mg
InChI formel InChI=1S/C9H6IN/c10-9-3-1-2-7-6-11-5-4-8(7)9/h1-6H
Formel vægt 254.9545