missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ DSPP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572040
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
DSPP Polyclonal Antibody for Western Blot, Flow
This gene encodes two principal proteins of the dentin extracellular matrix of the tooth. The preproprotein is secreted by odontoblasts and cleaved into dentin sialoprotein and dentin phosphoprotein. Dentin phosphoprotein is thought to be involved in the biomineralization process of dentin. Mutations in this gene have been associated with dentinogenesis imperfecta-1; in some individuals, dentinogenesis imperfecta occurs in combination with an autosomal dominant form of deafness. Allelic differences due to repeat polymorphisms have been found for this gene.
Tekniske data
| DSPP | |
| Polyclonal | |
| Unconjugated | |
| DSPP | |
| dentin matrix protein 3; dentin phosphophoryn; Dentin phosphoprotein; dentin phosphoryn; dentin sailophosphoprotein; dentin sialophosphoprotein; Dentin sialoprotein; Dentine sialoprotein; DFNA39; DGI1; Dmp2; Dmp3; DMP-3; Dpp; DSP; DSPP; DTDP2; RDSP2 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 1834 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9NZW4 | |
| DSPP | |
| KLH conjugated synthetic peptide between 47-76 amino acids of human DSPP. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion