Learn More
MP Biomedicals Drug of Abuse Rapid Test - Dip Card 10 parameters (AMP1000/BAR300/BUP10/BZD300/COC300/MTD300/M-AMP1000/OPI300/PCP25/THC50)
Product Code. 31454333 Shop All MP Biomedical Products
Change view
Click to view available options
Form:
Ready-to-Use (RTU)
Product Code. Form
31454333 Ready-to-Use (RTU)
1 options
Product Code. 31454333 Supplier MP Biomedicals Supplier No. 07RD707301
  • ONLINE EXCLUSIVE PRICE
    3230.00 DKK
    30.82 DKK / 1 tests
    Spare 3199.18 DKK
    99% Slukket
 Restricted Item
Forventet forsendelse: 30-12-2026
The Drug of Abuse Rapid Test - Dip Card 10 parameters is an immunochromatography based one step in vitro test.
The Drug of Abuse Rapid Test - Dip Card is based on the principle of specific immunochemical reaction between antibodies and antigen to analyze particular compounds in human urine specimen. The assay relies on the competition for the binding antibody. When drug is present in the urine specimen, it competes with drug conjugate for the limited amount of antibody-dye conjugate. When the amount of drug is equal or more than the cut-off, it will prevent the binding of drug conjugate to the antibody. Therefore, a positive urine specimen will not show a colored band on the test line zone, indicating a positive result, while the presence of a colored band indicates a negative result. A control line is present in the test window to work as procedural control. This colored band should always appear on the control line zone if the test device is stored in good condition and the test is performed appropriately.
  • CE-marked
  • It is designed for qualitative determination of drug substances and metabolites in human urine specimens
  • Tailor-made tests with customized combinations
  • More than 30 parameters: amphetamine, benzodiazepine, barbiturate, buprenorphine, cocaine, EDDP, ketamine, methamphetamine, MDMA, methadone, opiates, oxycodone, pencyclidine, THC, propoxyphene, tramadol, tricyclic antidepressant, methylphenidate, fentanyl, synthetic cannabis, clonazepam, cotinine, alcohol
  • Adulteration parameters: oxidants, bleach, pyridinium chlorochromate, nitrite, creatinine, pH and specific gravity
  • For this reference, the parameters and cut-off values are: Amphetamine 1000 ng/mL, Barbiturique 300 ng/mL, Buprenorphine 10 ng/mL, Benzodiazepine 300 ng/mL, Cocaine 300 ng/mL, Methadone 300 ng/mL, Methamphetamine 1000 ng/mL, Opiates 300 ng/mL, Phencyclidine 25 ng/mL, THC 50 ng/mL.
Specifications
CE Marker Self-declared
Clia Complexity Not Applicable
Detectable Analytes Amphetamine (AMP), Barbiturates (BAR), Buprenorphine (BUP), Benzodiazepines (BZO), Cocaine (COC), Methadone (MTD), Methamphetamine (mAMP), Opiates (OPI), Phencyclidine (PCP), Cannabinoids (THC)
Form Ready-to-Use (RTU)