Få mere at vide
Droperidol EP Impurity D (as Hydrochloride), Mikromol
Beskrivelse
Droperidol EP Impurity D (as Hydrochloride), Mikromol
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (1RS)-1-(4-(4-Fluorophenyl)-4-oxobutyl)-4-(2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)-1,2,3,6-tetrahydropyridine 1-Oxide Hydrochloride |
| CAS | 2724689-73-8 |
| Analyt- eller komponentnavne | (1RS)-1-(4-(4-Fluorophenyl)-4-oxobutyl)-4-(2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)-1,2,3,6-tetrahydropyridine 1-Oxide Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C22 H22 F N3 O3 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | (1RS)-1-[4-(4-Fluorophenyl)-4-oxobutyl]-4-(2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)-1,2,3,6-tetrahydropyridine 1-Oxide Hydrochloride, Droperidol Imp. D (EP), (1RS)-1-[4-(4-Fluorophenyl)-4-oxobutyl]-4-(2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)-1,2,3,6-tetrahydropyridine 1-Oxide Hydrochloride, Droperidol Imp. D (EP) as Hydrochloride, Droperidol Impurity D as Hydrochloride |
| InChI formel | InChI=1S/2C13H19NO3.H2O4S/c2*1-13(2,3)14-6-10-9(12(17)7-14)4-8(15)5-11(10)16;1-5(2,3)4/h2*4-5,12,15-17H,6-7H2,1-3H3;(H2,1,2,3,4) |
| Løsningstype | Neat |
| SMIL | Cl.[O-][N+]1(CCCC(=O)c2ccc(F)cc2)CCC(=CC1)N3C(=O)Nc4ccccc34 |
| Vis mere |